Order Entry
Poland
Orders LinkContactUsLinkComponent
Sodium L(+)-ascorbate 98%

Sodium L(+)-ascorbate 98%

Supplier:   Apollo Scientific
Sodium L(+)-ascorbate 98%
Supplier:  Apollo Scientific
Certificates
Sodium L(+)-ascorbate 98%

3 Options Available Below

Product Details & Documents
Synonyms:  Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
  • CAS Number:134-03-2
  • Formula:C₆H₇NaO₆
  • MDL Number:MFCD00082340
  • Molecular Weight:198.11 g/mol
  • Melting Pt:219…221 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 142 zł
    142 zł
    1 * 100 g
       142 zł

    Specifications:
    New Product
    New Product
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 354 zł
    354 zł
    1 * 500 g
       354 zł

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 637 zł
    637 zł
    1 * 1 kg
       637 zł

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.