Order Entry
Poland
ContactUsLinkComponent
Methyl cis-4-hydroxycyclohexanecarboxylate 95%
Methyl cis-4-hydroxycyclohexanecarboxylate 95%
Catalog # APOSOR922132-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl cis-4-hydroxycyclohexanecarboxylate 95%
Catalog # APOSOR922132-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :158.197 g/mol
Storage Temperature:Ambient
CAS Number:3618-03-9
Formula:C₈H₁₄O₃
MDL Number:MFCD20441962