Order Entry
Poland
ContactUsLinkComponent
cis-3,3,5-Trimethylcyclohexyl acetate 95%
cis-3,3,5-Trimethylcyclohexyl acetate 95%
Catalog # APOSOR916484-25G
Supplier:  Apollo Scientific
CAS Number:  
cis-3,3,5-Trimethylcyclohexyl acetate 95%
Catalog # APOSOR916484-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :184.28 g/mol
Storage Temperature:Ambient
CAS Number:24691-16-5
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00036661