Order Entry
Poland
ContactUsLinkComponent
2-(1-Pyrrolidinyl)-3-pyridinamine 95%
2-(1-Pyrrolidinyl)-3-pyridinamine 95%
Catalog # APOSOR913825-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(1-Pyrrolidinyl)-3-pyridinamine 95%
Catalog # APOSOR913825-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.22 g/mol
Storage Temperature:Ambient
CAS Number:5028-13-7
Formula:C₉H₁₃N₃
MDL Number:MFCD08700176