
(S)-(-)-N-Methoxymethyl-N-(trimethylsilyl)methyl-1-phenylethylamine
Catalog # APOSOR913190-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.45 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:143140-08-3
Formula:C₁₄H₂₅NOSi
MDL Number:MFCD06798960