Order Entry
Poland
Orders LinkContactUsLinkComponent
4,6-Dimethoxypyrimidine 97%

4,6-Dimethoxypyrimidine 97%

Supplier:   Apollo Scientific
4,6-Dimethoxypyrimidine 97%
Supplier:  Apollo Scientific
Certificates
4,6-Dimethoxypyrimidine 97%

6 Options Available Below

Product Details & Documents
  • CAS Number:5270-94-0
  • Formula:C₆H₈N₂O₂
  • MDL Number:MFCD00955844
  • Molecular Weight:140.14 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 107 zł
    107 zł
    1 * 250 mg
       107 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 121 zł
    121 zł
    1 * 1 g
       121 zł

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 149 zł
    149 zł
    1 * 5 g
       149 zł

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 276 zł
    276 zł
    1 * 25 g
       276 zł

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 080 zł
    1 080 zł
    1 * 100 g
       1 080 zł

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 5 240 zł
    5 240 zł
    1 * 500 g
       5 240 zł

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.