3,3'-Dihydroxybenzidine 97%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
- CAS Number:2373-98-0
- Formula:C₁₂H₁₂N₂O₂
- MDL Number:MFCD00039149
- Molecular Weight:216.24 g/mol
- Melting Pt:292 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 4,4'-Diamino-3,3'-dihydroxybiphenyl, 3,3'-Dihydroxy-4,4'-diaminobiphenyl, 4,4'-Diamino-[1,1'-biphenyl]-3,3'-diol, Benzidine-3,3'-diol
- CAS Number:2373-98-0
- Formula:C₁₂H₁₂N₂O₂
- MDL Number:MFCD00039149
- Molecular Weight:216.24 g/mol
- Melting Pt:292 °C
- Storage Temperature:Ambient



