
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:71989-18-9
:
Formula:C₂₄H₂₇NO₆
MDL Number:MFCD00037135
:425.48 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together