Order Entry
Poland
ContactUsLinkComponent
 
2-Hydroxy-3-methoxybenzaldehyde oxime 98%
Catalog # APOSOR911127-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Hydroxy-3-methoxybenzaldehyde oxime 98%
Catalog # APOSOR911127-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.17 g/mol
Storage Temperature:Ambient
CAS Number:2169-99-5
Formula:C₈H₉NO₃
MDL Number:MFCD01043210