2-(3-Methoxyphenyl)-2-methylpropanenitrile 96%
Catalog # APOSOR900540-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:175.23 g/mol
Storage Temperature:Ambient
CAS Number:17653-93-9
Formula:C₁₁H₁₃NO
MDL Number:MFCD09414739