4-(Cyclopropylamino)benzonitrile 98%
Catalog # APOSOR900312-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:158.20 g/mol
Storage Temperature:Ambient
CAS Number:1019607-55-6
Formula:C₁₀H₁₀N₂
MDL Number:MFCD11135501