2,4-Dibromopyridine 97%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:58530-53-3
- Formula:C₅H₃Br₂N
- MDL Number:MFCD01859720
- Molecular Weight:236.89 g/mol
- Melting Pt:35…40 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:58530-53-3
- Formula:C₅H₃Br₂N
- MDL Number:MFCD01859720
- Molecular Weight:236.89 g/mol
- Melting Pt:35…40 °C
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



