2-(1H-Indazol-6-yl)tetrahydro-1H-1λ~6~-isothiazole-1,1-dione
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:952183-42-5
:
Formula:C₁₀H₁₁N₃O₂S
MDL Number:MFCD09607932
:237.28 g/mol
Melting Pt:201…202 °C
Storage Temperature:Ambient
Frequently Bought Together