Order Entry
Poland
ContactUsLinkComponent
1-[4-(Methylsulphonyl)-2-nitrophenyl]-1,4-diazepane 95+%
1-[4-(Methylsulphonyl)-2-nitrophenyl]-1,4-diazepane 95+%
Catalog # APOSOR7795-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[4-(Methylsulphonyl)-2-nitrophenyl]-1,4-diazepane 95+%
Catalog # APOSOR7795-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.35 g/mol
Melting Pt:136…137 °C
Storage Temperature:Ambient
CAS Number:849035-89-8
Formula:C₁₂H₁₇N₃O₄S
MDL Number:MFCD04037963