Order Entry
Poland
Orders LinkContactUsLinkComponent
3,3'-Dimethylbiphenyl

3,3'-Dimethylbiphenyl

Irritant
Supplier:   Apollo Scientific
3,3'-Dimethylbiphenyl
Supplier:  Apollo Scientific
Certificates
3,3'-Dimethylbiphenyl

2 Options Available Below

Product Details & Documents
Synonyms:  3,3'-Bitolyl, 3,3'-Dimethyl-1,1'-biphenyl
  • CAS Number:612-75-9
  • Formula:C₁₄H₁₄
  • MDL Number:MFCD00008534
  • Molecular Weight:182.27 g/mol
  • Boiling Pt:286 °C/713mmHg
  • Melting Pt:44382 °C
  • Density:0.999 g/cm³ (25 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 191 zł
    191 zł
    1 * 1 g
       191 zł

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 573 zł
    573 zł
    1 * 5 g
       573 zł