
4-Chloropyridine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7379-35-3
:
Formula:C₅H₅Cl₂N
MDL Number:MFCD00012829
:150.01 g/mol
Melting Pt:210 °C (subl.)
Flash Pt:198 °C
EINECS:230-946-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together