
(4-Bromophenoxy)trimethylsilane 98%
Catalog # APOSOR6481-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:245.20 g/mol
Boiling Pt:105…107 °C/12mmHg
Density:1.268 g/cm³ (25 °C)
Flash Pt:97 °C
EINECS:241-831-0
:GHS07
Storage Temperature:Ambient
CAS Number:17878-44-3
Formula:C₉H₁₃BrOSi
MDL Number:MFCD00040713