Methyl-5-amino-2-(3-{[(tert-butoxy)carbonyl]amino}piperidin-1-yl)benzoate 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1280787-20-3
:
Formula:C₁₈H₂₇N₃O₄
MDL Number:MFCD18803532
:349.43 g/mol
Melting Pt:<50 °C
Storage Temperature:Ambient
Frequently Bought Together