Order Entry
Poland
ContactUsLinkComponent
Butyrophenone
Butyrophenone
Catalog # APOSOR60127-100G
Supplier:  Apollo Scientific
CAS Number:  
Butyrophenone
Catalog # APOSOR60127-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :148.2 g/mol
Boiling Pt:222…230 °C
Melting Pt:41579 °C
Density:1.021 g/cm³ (25 °C)
Flash Pt:88 °C
EINECS:207-799-7
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:495-40-9
Formula:C₆H₅C(O)(CH₂)₂CH₃
MDL Number:MFCD00009397