Order Entry
Poland
ContactUsLinkComponent
1-(4-Chlorophenylsulphonyl)-3,3-dimethyl-2-butanone 98%
Irritant
1-(4-Chlorophenylsulphonyl)-3,3-dimethyl-2-butanone 98%
Catalog # APOSOR6000-2.5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4-Chlorophenylsulphonyl)-3,3-dimethyl-2-butanone 98%
Catalog # APOSOR6000-2.5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

About this item

 :
 :274.77 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:207974-06-9
Formula:C₁₂H₁₅ClO₃S
MDL Number:MFCD00082685