Order Entry
Poland
Orders LinkContactUsLinkComponent
2,2-Dibromopropane

2,2-Dibromopropane

Bottle, 1 g
Catalog # APOSOR59903-1G
Irritant
Supplier:   Apollo Scientific
2,2-Dibromopropane
Catalog # APOSOR59903-1G
Supplier:  Apollo Scientific
CAS Number:  594-16-1
2,2-Dibromopropane

Bottle, 1 g

CAS Number:  594-16-1
Price
107 zł
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:594-16-1
  • Formula:H₃CBr₂CCH₃
  • MDL Number:MFCD00000119
  • Molecular Weight:201.89 g/mol
  • Boiling Pt:113…114 °C
  • Density:1.78 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:209-828-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications