Order Entry
Poland
ContactUsLinkComponent
tert-Butyl-3-(benzyloxy)-4-bromobenzoate 98%
tert-Butyl-3-(benzyloxy)-4-bromobenzoate 98%
Catalog # APOSOR5970-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-3-(benzyloxy)-4-bromobenzoate 98%
Catalog # APOSOR5970-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:247186-50-1
Formula:C₁₈H₁₉BrO₃
MDL Number:MFCD09027266