
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:104322-63-6
:
Formula:C₁₀H₁₅NO₃S
MDL Number:MFCD00075319
:229.29 g/mol
Melting Pt:172…174 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together