
4-Propylcyclohexanone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40649-36-3
:
Formula:C₉H₁₆O
MDL Number:MFCD00058690
Boiling Pt:60…62 °C
Flash Pt:86 °C
:140.23 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together