Order Entry
Poland
ContactUsLinkComponent
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 95%
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 95%
Catalog # APOSOR471615-100G
Supplier:  Apollo Scientific
CAS Number:  
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 95%
Catalog # APOSOR471615-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :341.28 g/mol
Storage Temperature:Ambient
CAS Number:32854-09-4
Formula:C₈H₁₈Cl₂N₂O₄S₂
MDL Number:MFCD00012490