3-(2-Chlorophenoxy)propionic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7170-45-8
:
Formula:C₉H₉ClO₃
MDL Number:MFCD02295728
:200.6190 g/mol
Storage Temperature:Ambient
Frequently Bought Together
1-Naphthol for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzoic acid methyl ester, EMPLURA®, Supelco®
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane, pure analytical reagent
Supplier: CHEMPUR
Chloroform stabilized with ethanol (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-DIMETHYLFORMAMIDE AR | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
2-Propanol 99.5%, gradient grade for HPLC
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific