Order Entry
Poland
ContactUsLinkComponent
Methyl-2,5-dibromobenzoate 98%
Irritant
Methyl-2,5-dibromobenzoate 98%
Catalog # APOSOR4690-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2,5-dibromobenzoate 98%
Catalog # APOSOR4690-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :293.94 g/mol
Melting Pt:48…51 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:57381-43-8
Formula:C₈H₆Br₂O₂
MDL Number:MFCD00144757