

(S)-(-)-1-Boc-3-aminopyrrolidine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:147081-44-5
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD03419271
:186.25 g/mol
Boiling Pt:216…217 °C
Density:1.067 g/cm³ (25 °C)
Flash Pt:91 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS05
:GHS06
Frequently Bought Together