Order Entry
Poland
ContactUsLinkComponent
2-(4-Nitrophenoxy)acetohydrazide 98%
Irritant
2-(4-Nitrophenoxy)acetohydrazide 98%
Catalog # APOSOR4176-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(4-Nitrophenoxy)acetohydrazide 98%
Catalog # APOSOR4176-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :211.18 g/mol
Melting Pt:184…186 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:75129-74-7
Formula:C₈H₉N₃O₄
MDL Number:MFCD00051973