
1-Amino-1-cyclopropanecarbonitrile hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:127946-77-4
:
Formula:C₄H₇ClN₂
MDL Number:MFCD04114063
:118.56 g/mol
Melting Pt:214…220 °C (dec.)
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
Frequently Bought Together