

2-Chloro-4-nitrophenyl isothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23165-64-2
:
Formula:C₇H₃ClN₂O₂S
MDL Number:MFCD00041244
:214.63 g/mol
UN:2923
ADR:8+6.1,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together