Order Entry
Poland
ContactUsLinkComponent
4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-carbaldehyde 95%
4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-carbaldehyde 95%
Catalog # APOSOR33485-1G
Supplier:  Apollo Scientific
CAS Number:  
4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-carbaldehyde 95%
Catalog # APOSOR33485-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.29 g/mol
Storage Temperature:Ambient
CAS Number:893737-04-7
Formula:C₁₆H₁₄O₃
MDL Number:MFCD06802589