1-(4-Methoxyphenyl)-3-methyl-1H-pyrazol-5-ylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:91331-86-1
:
Formula:C₁₁H₁₃N₃O
MDL Number:MFCD02091541
:203.25 g/mol
Melting Pt:104…105 °C
Storage Temperature:Ambient
Frequently Bought Together