
4,6-Dimethyl-2-pyrrolidin-1-ylnicotinonitrile
Catalog # APOSOR3254-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:201.27 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:693254-24-9
Formula:C₁₂H₁₅N₃
MDL Number:MFCD04209734