Order Entry
Poland
ContactUsLinkComponent
N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine 95%
N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine 95%
Catalog # APOSOR32532-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(4-Chloro-6-methoxy-2-pyrimidinyl)-N,N-dimethylamine 95%
Catalog # APOSOR32532-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.63 g/mol
Storage Temperature:Ambient
CAS Number:13012-26-5
Formula:C₇H₁₀ClN₃O
MDL Number:MFCD01935946