2,2-Dimethyl-4-(2-oxiranyl)tetrahydrofuro[3,4-d][1,3]dioxole 95%
Catalog # APOSOR32336-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.21 g/mol
Storage Temperature:Ambient
CAS Number:226709-43-9
Formula:C₉H₁₄O₄
MDL Number:MFCD03425721