

2-Methoxybiphenyl-3-isothiocyanate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:206761-68-4
:
Formula:C₁₄H₁₁NOS
MDL Number:MFCD00041078
Melting Pt:78…81 °C
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,III
:241.31 g/mol
:GHS05
:GHS07
Frequently Bought Together