Order Entry
Poland
ContactUsLinkComponent
5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole
5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole
Catalog # APOSOR32162-1G
Supplier:  Apollo Scientific
CAS Number:  
5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole
Catalog # APOSOR32162-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.24 g/mol
Storage Temperature:Ambient
CAS Number:321385-58-4
Formula:C₁₂H₁₀N₄
MDL Number:MFCD00138185