Order Entry
Poland
ContactUsLinkComponent
cis-3-Hydroxycyclobutanecarboxylic acid
cis-3-Hydroxycyclobutanecarboxylic acid
Catalog # APOSOR317124-1G
Supplier:  Apollo Scientific
CAS Number:  
cis-3-Hydroxycyclobutanecarboxylic acid
Catalog # APOSOR317124-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :116.1200 g/mol
Storage Temperature:Ambient
CAS Number:552849-33-9
Formula:C₅H₈O₃
MDL Number:MFCD12964956