
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) 98%
:27532-96-3
:
:C₆H₁₄ClNO₂
:MFCD00058255
:167.64 g/mol
:141…143 °C
:GHS07
:Ambient
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Diisopropyl ether 99+% stabilised, extra pure
: Thermo Fisher Scientific
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals