3,3-Dimethylcyclohexanone
New Product
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:2979-19-3
- Formula:C₈H₁₄O
- MDL Number:MFCD00040178
- Molecular Weight:126.2 g/mol
- Boiling Pt:175 °C
- Density:0.91 g/cm³ (25 °C)
- Flash Pt:59 °C
- Storage Temperature:Ambient
- UN:1224
- ADR:3,III
- :Density: 0.909 g/cm³ (25 °C)
5 Options Available Below
Product Details & Documents
- CAS Number:2979-19-3
- Formula:C₈H₁₄O
- MDL Number:MFCD00040178
- Molecular Weight:126.2 g/mol
- Boiling Pt:175 °C
- Density:0.91 g/cm³ (25 °C)
- Flash Pt:59 °C
- Storage Temperature:Ambient
- UN:1224
- ADR:3,III
- :Density: 0.909 g/cm³ (25 °C)
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



