5-Bromo-2-hydroxy-4-picoline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:164513-38-6
:
Formula:C₆H₆BrNO
MDL Number:MFCD03427664
:188.02 g/mol
Melting Pt:198…202 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together