
2,3-Dichloro-5-(trichloromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69045-83-6
:
Formula:C₆H₂Cl₅N
MDL Number:MFCD03701543
:265.3518 g/mol
Boiling Pt:278…279 °C
Density:1.636 g/cm³ (25 °C)
Flash Pt:65.56 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together