3,3-Dimethoxyoxetane
Supplier: Apollo Scientific
About this item
- CAS Number:922500-97-8
- Formula:C₅H₁₀O₃
- MDL Number:MFCD12545828
- Molecular Weight:118.13 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:922500-97-8
- Formula:C₅H₁₀O₃
- MDL Number:MFCD12545828
- Molecular Weight:118.13 g/mol
- Storage Temperature:Ambient
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.APOSOR312003-500MG - Specifications:
More
Specifications
Cat. No.APOSOR312003-1G - Specifications:
More
Specifications
Cat. No.APOSOR312003-5G
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



