Order Entry
Poland
ContactUsLinkComponent
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester
Irritant
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester
Catalog # APOSOR308114-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-(2-Aminospiro[3.3]hept-6-yl)carbamic acid-tert-butyl ester
Catalog # APOSOR308114-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.32 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1239589-52-6
Formula:C₁₂H₂₂N₂O₂
MDL Number:MFCD09054707