Order Entry
Poland
ContactUsLinkComponent
cis-3-(Boc-amino)cyclobutanemethanol
cis-3-(Boc-amino)cyclobutanemethanol
Catalog # APOSOR308096-1G
Supplier:  Apollo Scientific
CAS Number:  
cis-3-(Boc-amino)cyclobutanemethanol
Catalog # APOSOR308096-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.266 g/mol
Storage Temperature:Ambient
CAS Number:142733-64-0
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD08436074