
Bis(triphenylphosphine)palladium(II) chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13965-03-2
:
Formula:C₃₆H₃₀Cl₂P₂Pd
MDL Number:MFCD00009593
:701.91 g/mol
Melting Pt:310 °C (dec.)
EINECS:237-744-2
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together