2,7-Dibromocarbazole
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,7-Dibromo-9H-carbazole
- CAS Number:136630-39-2
- Formula:C₁₂H₇Br₂N
- MDL Number:MFCD09033507
- Molecular Weight:325.00 g/mol
- Melting Pt:225…230 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Synonyms: 2,7-Dibromo-9H-carbazole
- CAS Number:136630-39-2
- Formula:C₁₂H₇Br₂N
- MDL Number:MFCD09033507
- Molecular Weight:325.00 g/mol
- Melting Pt:225…230 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III



