Order Entry
Poland
ContactUsLinkComponent
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 95%
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 95%
Catalog # APOSOR304009-5G
Supplier:  Apollo Scientific
CAS Number:  
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 95%
Catalog # APOSOR304009-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :255.3600 g/mol
Melting Pt:90…92 °C
Storage Temperature:Ambient
CAS Number:86695-06-9
Formula:C₁₇H₂₁NO
MDL Number:MFCD01318247