Order Entry
Poland
ContactUsLinkComponent
4-tert-Butylphenyl isothiocyanate
Irritant
4-tert-Butylphenyl isothiocyanate
Catalog # APOSOR30261-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-tert-Butylphenyl isothiocyanate
Catalog # APOSOR30261-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :191.2926 g/mol
Boiling Pt:92…94 °C/0.5mmHg
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:19241-24-8
Formula:C₁₁H₁₃NS
MDL Number:MFCD00022059